Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CGB Rabbit Polyclonal Antibody

Supplier: BioVision
AB54410-100
10007-948EA 354.11 USD InStock
10007-948
Anti-CGB Rabbit Polyclonal Antibody
Antibodies
Human Chorionic Gonadotropin (hCG) consists of 237 amino acids, with chain-92 amino acids (α -subunit) and chain-145 amino acids (β-subunit).

Recommended Dilutions: Western Blot: 0.5-4 µg/ml;

Type: Primary
Antigen: CGB
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty