Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HCAM Rabbit Polyclonal Antibody

Supplier: BioVision
AB24504-100
10003-636EA 406.58 USD InStock
10003-636
Anti-HCAM Rabbit Polyclonal Antibody
Antibodies
Cell adhesion molecules (CAMs) are a family of closely related cell surface glycoproteins involved in cell-cell interactions during growth and are thoμght to play an important role in embryogenesis and development. The CAM family includes NCAM (neuronal cell adhesion molecule), ICAM (intracellular adhesion molecule), PECAM (platelet/endothelial cell adhesion molecule), BLCAM (produced by alternative splicing of the CD22 gene), VCAM (vascular cell adhesion molecule), and HCAM (homing cell adhesion molecule, also designed CD44), etc.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 20 µg/ml; and Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: HCAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty