Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SDC4 Rabbit Polyclonal Antibody

Supplier: BioVision
AB286154-100UG
10004-432EA 551.59 USD InStock
10004-432
Anti-SDC4 Rabbit Polyclonal Antibody
Antibodies
Syndecans are type I integral membrane proteoglycans that are involved in cell-extracellular matrix adhesion and growth factor binding. Syndecan-1 is an matrix receptor which binds to Collagens, Fibronectin and Thrombospondin. Syndecan-1 and Syndecan-3 interact with MK (midkine), a growth/differentiation factor involved in embryogenesis. Syndecan-2 is highly expressed at areas of high morphogenetic activity, such as epithelial-mesenchymal interfaces and the prechondrogenic and preosteogenic mesenchymal condensations. Syndecan-4 functions cooperatively with integrins in the processes of cell spreading and focal adhesion assembly.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 5-10 µg/ml; and Immunohistochemistry: 5 µg/ml(Frozen and paraffin sections)

Type: Primary
Antigen: SDC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty