Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BCL2L13 Rabbit Polyclonal Antibody

Supplier: BioVision
AB27795-100
10004-502EA 321.33 USD InStock
10004-502
Anti-BCL2L13 Rabbit Polyclonal Antibody
Antibodies
Bcl-rambo shares the common structural characteristics with other members of the anti-apoptotic Bcl-2 family members, but differs from them at its C-terminus, where a 250 amino acid sequence proceeds the membrance anchor region. It also differs from other pro-apoptotic Bcl-2 family members in that it membrane anchor C-terminus region is responsible for its apoptotic activity, not its Bcl-2 homology motifs.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BCL2L13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty