Would you like to log in again?
Autoclave Cleaning Brush Kit, Sklar
Supplier: Sklar
10-1424
94002-700EA 434.66 USD InStock
94002-700
Autoclave Cleaning Brush Kit, Sklar
Brushes
The Autoclave Cleaning Brush Kit contains one telescopic handle which extends from 61–122cm (24–48"), one large 13.9x19cm (5.5x7.5") cleaning brush with round corner bristles, and one hand brush with a 11.4cm (4.5") handle.
All components are reusable, nonsterile, and latex-free.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty