Would you like to log in again?
Autoclave Cleaning Brush, Sklar
Supplier: Sklar
10-1422
94002-698PK 276.54 USD InStock
94002-698
Autoclave Cleaning Brush, Sklar
Brushes
The Autoclave Cleaning Brush is designed for use on autoclave chambers and doors, sinks, and carts.
  • Nonsterile
  • Latex-free
The brush head measures 7.6x8.9cm (3x3.25"), and features white medical grade nylon bristles. Overall length is 48.9cm (19.25").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty