Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FCER2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: EBVCS-5]

Supplier: Tonbo Biosciences
50-0237-T100 50-0237-T500 50-0237-T025
10002-288EA 95.4 USD InStock
10002-288 10002-290 10002-292
Anti-FCER2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: EBVCS-5]
Antibodies
The EBVCS-5 antibody is specific for human CD23, also known as the low affinity IgE receptor (FceRII), a 45 kDa type II transmembrane protein and member of the immunoglobulin superfamily. It is expressed on most B cells and is upregulated upon activation. It is also present on mantle zone B cells, eosinophils, monocytes and a subset of T cells and platelets. CD23 plays a role in B cell development and differentiation, and also functions to regulate IgE production. Soluble secreted forms of CD23 have been reported to bind IgE, stimulate release of pro-inflammatory cytokines from monocytes, and play a role in B cell differentiation.

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: EBVCS-5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty