Would you like to log in again?
Ethanol Fuel from Corn Lab Activity
Supplier: Avantor
WARD470023-978
470023-978EA 172.99 USD InStock
470023-978
Ethanol Fuel from Corn Lab Activity
Educational Classroom Kits and Activities
First Hand Demonstration of Fuel Creation Processes
  • Compare and contrast two types of ethanol production methods and their respective combustion rates
  • Explore environmental, social, and economical impacts of ethanol as a fuel source
  • Includes materials for five setups or nine demonstrations
Demonstrate the creation of ethanol and examine its use and effectiveness with this student experiment. By performing germination, fermentation, and distillation procedures, students will produce ethanol and other products. Two methods are offered for germinating the corn seed and acquiring the enzymes required for saccharification of starches in the corn, so that students can compare each process. Applying concepts of volatility of liquids, evaporation, and condensation, students will examine the combustion of both lab-grade ethanol and corn ethanol. Observing the differences and similarities between the ethanol types will establish a complete understanding of this fuel. Students will then discuss the future of this new fuel source and its environmental, economic, and social impacts. Includes materials for five lab setups.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty