Would you like to log in again?
Herbarium Mounting Cards
Supplier: XPEDX
WHC1SCAR
470195-076PK 19.25 USD InStock
470195-076
Herbarium Mounting Cards
Educational Materials Environmental Educational Materials Environmental Learning Activities
These simple white cards are made of 0.021" thick, 5-ply paper and can be used for mounting frequently handled specimens so they won't be destroyed.
  • Size: 11.5×16.5"
The cards will not yellow under normal conditions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty