Would you like to log in again?
Schaedler Agar with Sheep Blood, Hemin and Vitamin K, Hardy Diagnostics
Supplier: Hardy Diagnostics
A98 A98
10753-984PK 125.52 USD InStock
10753-984 CA10753-984
Schaedler Agar with Sheep Blood, Hemin and Vitamin K, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Schaedler agar with 5% sheep blood, Hemin and Vitamin K, is used in the cultivation of anaerobic bacteria.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty