Would you like to log in again?
Columbia CNA Agar with Sheep Blood, Hardy Diagnostics
Supplier: Hardy Diagnostics
GA50 GA50
10753-962PK 36.43 USD InStock
10753-962 CA10753-962
Columbia CNA Agar with Sheep Blood, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Columbia CNA (Colistin and Nalidixic Acid) agar with 5% sheep blood, for the selective isolation of Gram- positive bacteria, RSR, reduced stacking ring for automated inoculating devices.
Recommended for use as a selective growth medium for the isolation and differentiation of Gram- positive cocci from clinical and non-clinical specimens with mixed flora.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty