Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Classic++™ Taq DNA Polymerase, Tonbo Biosciences
Supplier: Tonbo Biosciences
Classic++™
31-5000-2000U 31-5000-0500U
10002-198EA 600 USD InStock
10002-198 10002-196
Classic++™ Taq DNA Polymerase, Tonbo Biosciences
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Classic++ Taq DNA Polymerase is a robust next generation ecombinant Taq polymerase, optimized for exceptional everyday PCR
For added convenience and value, the advanced Reaction Buffer (5X) includes dNTPs and MgCl2. Use Classic++ Taq DNA Polymerase for high-throughput PCR, TA cloning, genotyping, library construction and RT-PCR under fast or standard cycling conditions.
Works well with crude samples and complex template DNA.
Convenient and reliable, Classic++ Taq polymerase is suitable for routine PCR protocols and has been engineered to provide enhanced speed, yield and specificity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty