Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCND2 Mouse Monoclonal Antibody [clone: CY-D2]

Supplier: Rockland Immunochemicals
200-301-P32
10801-046EA 528 USD InStock
10801-046
Anti-CCND2 Mouse Monoclonal Antibody [clone: CY-D2]
Antibodies
Inaba et al. (1992) used murine cDNA clones for 3 cyclin D genes that are normally expressed during the G1 phase of the cell cycle to clone the cognate human genes. D-type cyclins (cyclins D1, D2, and D3) are regarded as essential links between cell environment and the core cell cycle machinery. cyclin D2(CCND2) gene ia assigned to 12p13. This antibody is suitable for researchers interested in cancer research, autoimmune disease, neurodegenerative disease, MAP Kinase Signaling, AKT Signaling, Ubiquitin research, and Cell Cycle research.

Recommended Dilutions: ELISA: 1:40,000; Immunohistochemsitry: 1:100-1:500; Western Blot: 2-4µg/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: CCND2
Clonality: Monoclonal
Clone: CY-D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty