Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GAB1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-G66S 200-401-G66S
10800-032EA 148 USD InStock
10800-032 CA10800-032
Anti-GAB1 Rabbit Polyclonal Antibody
Antibodies
GAB1 antibody detects GAB1 which is a member of the IRS1-like multisubstrate docking protein family. The protein is an important mediator of branching tubulogenesis and plays a central role in cellular growth response, transformation and apoptosis. Two transcript variants encoding different isoforms have been found for this gene. GAB1 plays a role in intracellular signaling cascades triggered by activated receptor-type kinases. It is known to play a role in FGFR1 signaling and is probably involved in signaling by the epidermal growth factor receptor (EGFR) and the insulin receptor (INSR). GAB1 interacts with GRB2 and with other SH2-containing proteins. It is known to interact with phosphorylated LAT2, PTPRJ, FRS2, GRB2, PIK3R1 and SOS1. GAB1 gets phosphorylated in response to FGFR1 activation. This tyrosine phosphorylation of GAB1 mediates interaction with several proteins that contain SH2 domains. Anti-GAB1 Antibody is ideal for investigators involved in Cell Signaling, Cancer, Neuroscience and Signal Transduction research.

Recommended Dilutions: ELISA: 1:2,000 - 1:10,000; Western Blot: 1:500-1:2000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: GAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty