Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EZH2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-W53
10802-658EA 518 USD InStock
10802-658
Anti-EZH2 Rabbit Polyclonal Antibody
Antibodies
EZH2 is a histone-lysine methyltransferase which methylates ‘Lys-9’ and ‘Lys-27’ of histone H3, leading to transcriptional repression. It is a member of the polycomb group (PcG) family which form multimeric protein complexes and are involved in maintaining the transcriptional repressive state of genes over successive cell generations. The EZH2 activity is dependent on the association with other components of the PRC2 complex (EED, EZH2, SUZ12/JJAZ1, RBBP4 and RBBP7). EZH2 may play a role in the hematopoietic and internal nervous systems. Over-expression of EZH2 is observed during advanced stages of prostate cancer and breast cancer. Anti-EZH2 Antibody is ideal for research in Epigenetics, Gene Expression, Cell Biology and Cancer.

Recommended Dilutions: ChIP: 2.5 ?g per IP; Immunohistochemsitry: 1:100 - 1:500; Western Blot: 1:1,000; contains 0.05% (w/v) Sodium Azide and 0.05% ProClin 300

Type: Primary
Antigen: EZH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty