Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TBC1D10C Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AB2
10800-518EA 518 USD InStock
10800-518
Anti-TBC1D10C Rabbit Polyclonal Antibody
Antibodies
Antigen binding by the T-cell receptor (TCR) is one of the critical first steps in the immune response, triggering a cascade of signaling pathways that ultimately lead to T-cell activation. Screening a yeast two-hybrid screen of a human T-cell cDNA library with calcineurin, a protein phosphatase involved in multiple signaling pathways including T-cell activation, resulted in the identification of Carabin, a member of the TBC1 domain family of proteins, as a calcineurin-binding protein. Unlike other members of the TBC1 domain protein family which are thought to have a role in regulating cell growth and differentiation, further experiments demonstrated that Carabin is part of a negative regulatory loop for the intracellular TCR signaling pathway as well as an inhibitor of the Ras signaling pathway, suggesting that Carabin may also mediate crosstalk between calcineurin and Ras. Carabin antibody does not recognize TBC1D10A or TBC1D10B. Carabin is known to exist in multiple isoforms.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 2.5 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: TBC1D10C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty