Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-COMMD7 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AM5
10800-868EA 518 USD InStock
10800-868
Anti-COMMD7 Rabbit Polyclonal Antibody
Antibodies
COMMD7 (COMM domain-containing protein 7) plays a role in the regulation NF-kappaB (1). It associates with the NF-kB complex and suppresses its transcriptional activity (2). COMMD7 contains 1 COMM domain and widely expressed with highest expression in lung (3). COMMD7 is a newly identified gene overexpressed in hepatocellular carcinoma (HCC) (4). COMMD7 is associated with tumor invasion and poor prognosis by NF-kappaB signaling pathway (4).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 µg/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: COMMD7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty