Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DENR Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AU9
10802-436EA 518 USD InStock
10802-436
Anti-DENR Rabbit Polyclonal Antibody
Antibodies
DENR (density-regulated protein), also designated DRP or smooth muscle cell associated protein 3 (SMAP-3), is a 198 amino acid protein that may be involved in the translation of target mRNAs by scanning and recognition of the initiation codon (1). It plays a role in the modulation of the translational profile of a subset of cancer-related mRNAs when recruited to the translational initiation complex by the oncogene MCTS1 (2,3). DENR contains a SUI1 domain and is found in a variety of tissues with highest levels present in skeletal and cardiac muscle. It is up-regulated in ovarian and breast cancer cells by ERBB2 overexpression (4).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DENR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty