Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FOXO4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BG8
10801-472EA 518 USD InStock
10801-472
Anti-FOXO4 Rabbit Polyclonal Antibody
Antibodies
FOXO4 is a ubiquitously expressed protein member of a subfamily of the forkhead homeotic gene family of transcription factors and shuttles between the cytoplasm and nucleus. FOXO transcription factors are key players of cell fate decisions, metabolism, stress resistance, tumor suppression and are regulated by growth factors, oxidative stress or nutrient deprivation. In the absence of PI3K/AKT activation, FOXO4 localizes in the nucleus where it functions as a transcription factor. FOXO4 can also be phosphorylated by JNK following induction of reactive oxygen species (ROS), resulting in transcriptional activation and the induction of a negative feedback mechanism to counteract the ROS. It is through this mechanism that FOXO4 is thought to sensitize cancer cells to doxorubicin-mediated toxicity.

Recommended Dilutions: ELISA: 1:5000-1:10000; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 0.5-1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: FOXO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty