Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ANXA5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-P51 600-401-P51
10801-854EA 487 USD InStock
CA10801-854 10801-854
Anti-ANXA5 Rabbit Polyclonal Antibody
Antibodies
Annexin V also know as endonnexin II (ENX2), or placental protein 4 (PP4). Endonexin II is a member of the family of Ca(2+)-dependent phospholipid binding proteins, known as annexins. It bind to the phospholipids that are preferentially located on the cytosolic face of the plasma membrane. It has a relative molecular weight of about 35,000. The gene lies on mouse chromosome 3 in close linkage with the fibroblast growth factor 2 (basic) gene and is syntenic with other genes known to have orthologous counterparts on human chromosome 4q. The PP4 cDNA encoded a protein of 320 amino acid residues. A single mRNA, approximately 1.6 kb long, was found to be expressed in human cell lines and placenta. PP4 is an anticoagulant protein that acts as an indirect inhibitor of the thromboplastin-specific complex, which is involved in the blood coagulation cascade. This antibody is suitable for researchers interested in cell signaling and neuroscience research.

Recommended Dilutions: Immunohistochemsitry: 1:100-1:500; Western Blot: 0.5µg/mL; 0.05mg Thimerosal

Type: Primary
Antigen: ANXA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty