Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AR Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-W93 600-401-W93
10800-014EA 518 USD InStock
CA10800-014 10800-014
Anti-AR Rabbit Polyclonal Antibody
Antibodies
The androgen receptor (AR) is a DNA-binding transcription factor that regulates genes critical for the development and maintenance of the male sexual phenotype. Defects in androgen receptor have been shown to play a role in prostate cancer, and inhibition of AR activity through modulation of signal transduction pathways may delay prostate cancer progression. Multiple phosphorylation sites have been identified on the androgen receptor that affect cross-talk between growth factor signaling and androgen in prostate development and cancer. One of these sites, at Ser94, appears constitutively phosphorylated and exhibits no response to treatments with stimulating hormone. The site at Ser94 is unique among the AR phosphorylation sites in that it does not achieve a maximal level of phosphorylation between translation and the initial round of nuclear import, having a strong bias for androgen-independent phosphorylation in the cytoplasm. Androgen Receptor pS94 Antibody is ideal for researchers interested in cancer research.

Recommended Dilutions: ELISA: 1:10,000; Western Blot: 1:1000;

Type: Primary
Antigen: AR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Non-Human Primates

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty