Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RUNX1T1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X43 600-401-X43
10799-996EA 487 USD InStock
10799-996 CA10799-996
Anti-RUNX1T1 Rabbit Polyclonal Antibody
Antibodies
RUNX1T1 is a putative transcription factor which forms a heterodimer with CBFA2T3. Defects in RUNX1T1 have been associated with acute myeloid leukemia (AML-M2) and may be a cause of colorectal cancer. Anti-AML-ETO Antibody is ideal for research in Gene Expression and Cancer.

Recommended Dilutions: ELISA: 1:100; Western Blot: 1:1,000; contains 0.05% (w/v) Sodium Azide and 0.05% ProClin 300

Type: Primary
Antigen: RUNX1T1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty