Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AGTR2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X82 600-401-X82
10799-602EA 518 USD InStock
CA10799-602 10799-602
Anti-AGTR2 Rabbit Polyclonal Antibody
Antibodies
Angiotensin II is a potent vasopressor hormone and a primary regulator of aldosterone secretion that acts through at least two types of receptors, AGTR1 and AGTR2. It is an important effector controlling blood pressure and volume in the cardiovascular system and plays a major role in the development of the mammalian kidney and urinary tract. Like AGTR1, AGTR2 is a seven transmembrane G protein-coupled receptor (GPCR), but AGTR2 does not demonstrate most of the classic features of GPCR signaling. AGTR2 is involved in a wide range of activities, including the induction of neurite outgrowth and the inhibition of cellular proliferation, in addition to the known function of mediation of vasoconstriction.

Recommended Dilutions: ELISA: 1:5,000; Immunohistochemsitry: 5 ?g/mL; Western Blot: 0.5 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: AGTR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty