Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AMIGO1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Y10 600-401-Y10
10799-992EA 518 USD InStock
10799-992 CA10799-992
Anti-AMIGO1 Rabbit Polyclonal Antibody
Antibodies
The amphoterin-induced gene and ORF (AMIGO1) protein is a brain-enriched, glycosylated transmembrane immunoglobulin (Ig) superfamily protein with six extracellular leucine-rich repeats (LRRs) and one Ig-like domain. It and the related proteins AMIGO2 and AMIGO3 are thought to be cell adhesion molecules expressed on fiber tracts of neuronal tissues and participate in their formation (1). AMIGO1 has also been suggested to play important roles in dendritic outgrowth during development and could modulate the survival of developing and adult neurons (2).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 - 2 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: AMIGO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty