Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ANGPTL3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Y15 600-401-Y15
10800-020EA 523 USD InStock
10800-020 CA10800-020
Anti-ANGPTL3 Rabbit Polyclonal Antibody
Antibodies
The angiopoietin-related protein 3 (ANGPTL3) is a member of a family of secreted proteins that function in angiogenesis (1). ANGPTL3 is processed into an N-terminal coiled-coil domain-containing chain and a C-terminal fibrinogen chain. The N-terminal chain is important for lipid metablism, while the C-terminal chain may be involved in angiogenesis (2,3). Recent experiments have shown that a deficiency of ANGPTL3 is associated with increased insulin sensitivity, lipoprotein lipase activity, and decreased serum free fatty acids, suggesting that ANGPTL3 may also play a role in modulating glucose metabolism (4).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 - 2 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: ANGPTL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty