Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AQP2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Y54 600-401-Y54
10801-892EA 530 USD InStock
10801-892 CA10801-892
Anti-AQP2 Rabbit Polyclonal Antibody
Antibodies
Aquaporins are membrane proteins that serve in the transfer of water and small solutes across cellular membranes. One such aquaporin, aquaporin-2 (AQP2) is located in the kidney collecting tubule and plays a critical role in water reabsorbtion (1). AQP2 is mainly localized in intracellular vesicles but upon stimulation with anti-diuretic hormone (ADH), AQP2 is translocated to the apical plasma membrane by exocytic fusion of AQP2-bearing vesicles (2). Mutations in this gene have been linked to autosomal dominant and recessive forms of nephrogenic diabetes insipidus (3).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 - 2 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: AQP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty