Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NKX3-2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Z34
10802-012EA 518 USD InStock
10802-012
Anti-NKX3-2 Rabbit Polyclonal Antibody
Antibodies
BAPX1 is the mammalian homolog of the Drosophila bagpipe homeobox gene and is expressed in the splanchnic mesoderm and embryonic skeleton. It is one of the earliest developmental markers for the sclerotome portion of the somite and the gut mesentery. BAPX1 is required for normal skeletal development; homozygous inactivating mutations in the BAPX1 gene result in spodylo-megaepiphyseal-metaphyseal dysplasia (SMMD). It has also been suggested to play a role in the proper development of the mammalian gut and is required for distal stomach development as part of a BARX1-dependent pathway.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 µg/mL; Immunofluorescence Microscopy: 20 µg/mL; Western Blot: 1 - 2 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: NKX3-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty