Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Epitope Tag Antibody Sampler Kit

Supplier: Rockland Immunochemicals
K915
RLK915EA 611 USD InStock
RLK915
Epitope Tag Antibody Sampler Kit
Protein Linking and Labeling Systems
Epitope tag antibody sampler kit detects overexpressed fusion proteins containing the corresponding epitope tags. DYKDDDDK Tag Antibody recognizes the DYKDDDDK peptide (the same epitope recognized by Sigma's Anti-FLAG® antibodies). Epitope tags are useful for the labeling and detection of proteins using western blotting, immunoprecipitation, and immunostaining techniques.

Contains: 6X HIS antibody, GFP Antibody, DYKDDDDK Antibody (Binds to same epitope as Sigma's Anti-FLAG® M2 Antibody), GST Antibody and Conjugated Secondary Antibodies

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty