Would you like to log in again?
DURAN® PURE Laboratory Bottles, DWK Life Sciences
Supplier: DWK Life Sciences
818013608
10796-642CS 153.26 USD InStock
10796-642
DURAN® PURE Laboratory Bottles, DWK Life Sciences
Bottles Glass Bottles
DURAN PURE is a complete, USP compliant range of bottles from 25 ml to 20 L.
Bottles are made from Type 3.3 borosilicate glass. A protective dust cover is applied after the bottle is cooled on the production line. The bottles are packaged in shrinked-wrapped modules.
DURAN PURE Premium screw caps are manufactured from fluoropolymer (PFA-TpCh260TZ). The caps are colorant and pigment free. They are suitable formost sterilization methods.
These validation ready sizes are scalable from pilot to production volumes, with the added durability to go from hot 'sterile' fills to cold storage. These bottles meet the documentation and production requirements of national and international pharmacopeias for primary packaging materials.
Certifications: DURAN PURE Bottles are supported by a comprehensive, easy to access online validation document package and support services. The documentation package includes a certificate of lot analysis, Type III Drug Master File, and Qualification documentation.
Delivery information: Screw caps are sold separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty