Would you like to log in again?
Shear Power™ Spring Action Shears, Emergency Medical International
Supplier: Emergency Medical International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
2240
10755-988EA 41.3 USD InStock
10755-988
Shear Power™ Spring Action Shears, Emergency Medical International
Scissors
Shear Power™ has an angled hardened stainless steel blade with a comfortable spring load non-slip rubber handle
The spring action in the shears increases torque without straining your hands. It can easily cut though wire, cable, leather, webbing, etc. Shear Power™ has thumb operated lock to ensure that the blades are not exposed, and that the handles lock for safety when stored or when not in use. Weight 4 oz.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty