Would you like to log in again?
Grp78/BiP ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
900-214-0001
10014-512EA 969.11 USD InStock
10014-512
Grp78/BiP ELISA Kit, Enzo Life Sciences
Assays ELISAs
High-sensitivity ELISA kit for quantifying both normal and upregulated levels of Grp78/BiP for unfolded protein response, cancer and neurodegenerative disease research.
For the quantitative determination of human, mouse and rat Grp78/BiP in cell lysate and serum samples.
Ordering information: Contains microtiter plate, conjugate, antibody, assay buffer, wash buffer concentrate, standard, TMB substrate, stop solution 2, extraction reagent.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty