Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 0.9% Physiological saline endotoxin-free, sterile

Supplier: Enzo Life Sciences
Synonyms: SaltHaliteSea saltRock saltSaline

505-009-LD15
10014-564EA 63.62 USD InStock
10014-564
Sodium chloride 0.9% Physiological saline endotoxin-free, sterile
Sodium chloride
Sterile and endotoxin-free physiological saline for use with TLRgrade™ or endotoxin-free grade reagents.

Product has been subjected to multiple rounds of LPS removal by adsorption with activated charcoal.


Formula: NaCl
MW: 58.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now
Specification Test Results
Endotoxin content 0.0002 EU/µg (pyrotell gel-clot endotoxin test)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty