Would you like to log in again?
Whizard® Defender 13 Gloves, Wells Lamont
Supplier: Wells Lamont
135472 135476 135473 135474 135475
89015-549EA 28.38 USD InStock
89015-549 89015-543 89015-541 89015-547 89015-545
Whizard® Defender 13 Gloves, Wells Lamont
Gloves
The prewashed and preshrunk gloves feature Honeywell Spectra® Fiber.
  • Prewashed and Preshrunk
  • Spectra Fiber
  • Antimicrobial Fibers
  • Stainless Steel
  • High Abrasion Resistance
The combination of durable fibers and stainless steel provides strength and offers high abrasion resistance
The ambidextrous gloves are ideal for light-duty applications.
Antimicrobial fibers offer an additional layer of protection by inhibiting bacterial and fungal growth.
Certifications: ANSI Cut Level 4, A5. CFR 21 approved for food contact.
Ordering information: The gloves are sold in multiples of six pieces.
Caution: Caution Information: Cut-resistant gloves are extremely cut resistant but not cut proof. Do not subject to high speed or highly serrated blades. Always disconnect power before cleaning or removing slicer blades.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty