Would you like to log in again?
Whizard® Handguard II White Gloves, Wells Lamont
Supplier: Wells Lamont
333025 333027 333019 333021 333023 134147
32935-139EA 50.38 USD InStock
32935-139 87000-551 32935-179 32935-143 32935-141 32935-183
Whizard® Handguard II White Gloves, Wells Lamont
Gloves
These heavy-duty, cut-resistant gloves feature Honeywell Spectra® Fiber.
  • Prewashed and Preshrunk
  • Honeywell Spectra Fiber
  • Cut Resistant
The combination of high-strength fibers and stainless steel provides exceptional protection against cuts and slashes.
They work best in meat, poultry, and seafood processing.
The prewashed, preshrunk, and ambidextrous gloves have a 3" cuff for wrist protection.
Certifications: ANSI Cut Level 5. CFR 21 approved for food contact.
Ordering information: The gloves are sold in multiples of six pieces.
Caution: Caution Information: Cut-resistant gloves are extremely cut resistant but not cut proof. Do not subject to high speed or highly serrated blades. Always disconnect power before cleaning or removing slicer blades.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty