Would you like to log in again?
JACKSON SAFETY® V60 Safeview™ Safety Eyewear with RX Inserts, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
JACKSON SAFETY®
38504 38507 38505 38503
10118-250CS 106.42 USD InStock
10118-250 10118-220 10118-128 10118-126
JACKSON SAFETY® V60 Safeview™ Safety Eyewear with RX Inserts, KIMBERLY-CLARK PROFESSIONAL®
Safety Glasses
JACKSON SAFETY® V60 Safeview Safety Eyewear with RX Inserts provide a convenient way to get prescription eyewear at a local vision center with the included prescription insert which clicks easily into polycarbonate frame.
  • Clear peripheral vision without the need for side shields
  • Lightweight safety eyewear with adjustable ratchet-action temples for a custom fit
  • Corrective lenses are hard coated for scratch resistance and longer life
These safety glasses have polycarbonate lenses that help protect the prescription insert from damage and provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss.
Both men and women will find these stylish glasses to be comfortable and functional for construction, manufacturing, and a host of other industries.
Base curve 6.5
Certifications: Comply with the ANSI / ISEA Z87.1+ standard for personal eye protection.
Ordering information: Every pair includes a neck cord.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty