Would you like to log in again?
Vacuum Gauges, Digital, Chemglass
Supplier: Chemglass
AF-0301-B-01
89028-168EA 2061.33 USD InStock
89028-168
Vacuum Gauges, Digital, Chemglass
Pumps Vacuum Gauges
Digital vacuum gauge has an analog style screen for easy viewing.
  • User selected units of measure
The transducer is made of corrosion resistant ceramic for durability and measures absolute pressures. Powered by a 9 volt lithium battery with adjustable sleep timer for long battery life.
Delivery information: Supplied with battery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty