Would you like to log in again?
AMPIGENE® qPCR Green Mix Hi-ROX, Enzo Life Sciences
Supplier: Enzo Life Sciences
AMPIGENE®
NUC104-0200 NUC104-1000
10014-634EA 155.49 USD InStock
10014-634 10014-506
AMPIGENE® qPCR Green Mix Hi-ROX, Enzo Life Sciences
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Optimized dye-based qPCR.
AMPIGENE® qPCR Green Mixes use the latest developments in polymerase technology and buffer chemistry to deliver high performing dye-based real-time PCR with minimal optimization. AMPIGENE® qPCR Green Mixes use a proprietary intercalating dye that does not inhibit PCR.An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity. ROX is a reference dye which is used to normalize fluorescent signal in qPCR. AMPIGENE® qPCR Green Mixes come with either Lo-ROX or Hi-ROX concentration. For Lo-ROX, see Prod. No. ENZ-NUC103. Choose the appropriate ROX concentration for the instrument.
Ordering information: Contains 2x AMPIGENE® qPCR Green Mix Hi-ROX (includes dNTPs and polymerases).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty