Would you like to log in again?
AMPIGENE® qPCR Probe Mix No-ROX, Enzo Life Sciences
Supplier: Enzo Life Sciences
AMPIGENE®
NUC107-0200 NUC107-1000
10014-656EA 201.82 USD InStock
10014-656 10014-658
AMPIGENE® qPCR Probe Mix No-ROX, Enzo Life Sciences
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Optimized probe-based qPCR.
AMPIGENE® qPCR Probe Mixes use the latest developments in polymerase technology and buffer chemistry to deliver high performing probe-based real-time PCR with minimal optimization. AMPIGENE® qPCR Probe Mixes are designed for use on a variety of probes including: Taqman ®, Molecular Beacons ®, and Scorpion probes ®. An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity. ROX is a reference dye which is used to normalize fluorescent signal in qPCR. AMPIGENE® qPCR Probe Mixes come with either Lo-ROX, Hi-ROX, or No-ROX concentration. For Lo-ROX and Hi-ROX, see Prod. Nos. ENZ-NUC105 and ENZ-NUC106. Choose the appropriate ROX concentration for the instrument (see instrument compatibility for reference dye chart below.)
Ordering information: Contains 2x AMPIGENE® qPCR Probe Mix NO-ROX (includes dNTPs and polymerases).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty