Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-3-fluorobenzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Amino-3-fluorophenylboronic acid pinacol ester2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anilineBenzenamine, 2-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-

H62033-03 H62033-MD
AAH62033-MDEA 167.99 USD InStock
AAH62033-MD AAH62033-03
2-Amino-3-fluorobenzeneboronic acid pinacol ester 96%
2-Amino-3-fluorobenzeneboronic acid pinacol ester


Formula: C₁₂H₁₇BFNO₂
MW: 237.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025468
CAS Number: 1310384-26-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty