Would you like to log in again?
HyFlex® 11-531 Ultralight Weight, 18-Gauge, Light Duty Gloves, Palm Dipped, Ansell
Supplier: Ansell
HyFlex®
11531070 11531090 11531110 11531100 11531060 11531080
10001-986CS 2900.82 USD InStock
10001-986 10001-988 10001-994 10001-984 10001-990 10001-992
HyFlex® 11-531 Ultralight Weight, 18-Gauge, Light Duty Gloves, Palm Dipped, Ansell
Gloves
Offering exceptional grip performance and palm dip protection in a comfortable, ultralight weight, cut-resistant glove, the HyFlex® 11-531 epitomizes the convergence of protection and performance.
  • Ergonomic design for comfortable movement
  • Great breathability
  • Superior tactile sensitivity
  • Excellent abrasion resistance
  • Strong grip
It features an 18G ultra-lightweight liner with INTERCEPT TECHNOLOGY® yarns that provides EN Cut Level 3 (ANSI Cut Level 2) protection.
This glove's ultralight weight enables unparalleled dexterity and refined mobility for bare hand-like tactility. The ZONZ™ comfort fit technology uses selected yarns and varying stitch designs to tailor overall glove fit to enhance hand movement for higher dexterity and reduced fatigue. Its silicone-free design is paint- and finish-process-friendly and dirt-masking palm allows longer wear times.
INTERCEPT TECHNOLOGY® yarn has been designed to perform at or exceed industry standards. This Ansell proprietary technology offers best-in-class cut protection and increased comfort with dexterity. Palm coated protection provides grip while not limiting comfort, and back-of-hand breathability. ANSELL GRIP™ technology is a coating treatment that minimizes the force required to grip oily or wet tools or materials, reducing hand and arm fatigue and improving control, safety and productivity. The HyFlex® 11-531 glove also features FORTIX™ technology which applies a thin, resilient and breathable nitrile foam coating, which extends gloves’ working life and improves comfort in abrasive applications.
For use in the aerospace, automotive, machinery and equipment, and metal fabrication industries for assembly of small sharp parts, handling small sharp metal parts, light-duty stamping and maintenance.
Certifications: ANSI Cut Level 2. ANSI Abrasion Level 4.
Packaging: Packaged 12 pairs in a bag; 12 bags in a carton.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty