Would you like to log in again?
HyFlex® 11-537 Ultralight Weight, 18-Gauge Gloves, 3/4 Dipped, Ansell
Supplier: Ansell
HyFlex®
11537090 11537110 11537080 11537060 11537070 11537080 11537100
10001-998CS 3222.68 USD InStock
10001-998 10002-002 10002-068 10002-070 CA10002-072 10002-072 10002-000
HyFlex® 11-537 Ultralight Weight, 18-Gauge Gloves, 3/4 Dipped, Ansell
Gloves
Offering exceptional grip performance and ³/₄ dip protection in a comfortable, ultralight weight, cut-resistant glove, the HyFlex® 11-537 epitomizes the convergence of protection and performance.
  • Great cut resistance
  • Unparalleled dexterity
  • Superior tactile sensitivity
  • Reduced hand fatigue
It features an 18G ultra-lightweight liner with INTERCEPT TECHNOLOGY® yarns that provides EN Cut Level 3 (ANSI Cut Level 2) protection.
New-to-the-world INTERCEPT TECHNOLOGY® yarn has been designed to perform at or exceed industry standards. This Ansell proprietary technology offers best-in-class cut protection and increased comfort with dexterity. The 11-537 features an extended ³/₄ coating geometry on back-of-hand for comprehensive protection against snags and mechanical aggression. ANSELL GRIP™ technology is a coating treatment that minimizes the force required to grip oily or wet tools or materials, reducing hand and arm fatigue and improving control, safety and productivity. The HyFlex® 11-537 glove also features FORTIX™ technology which applies a thin, resilient and breathable nitrile foam coating, which extends gloves’ working life and improves comfort in abrasive applications.
For use in the aerospace, automotive, machinery and equipment, and metal fabrication industries for assembly of small sharp parts, handling small sharp metal parts, light-duty stamping and maintenance.
The liner also features FORTIX™ technology, providing EN Abrasion Level 4 (ANSI Abrasion Level 4) protection in a comfortable, flexible ultralight weight style. The gloves are a game changer for applications where workers have traditionally guarded against cut risks with heavier styles that limit their dexterity and breathability. This glove's ultralight weight enables unparalleled dexterity and refined mobility for barehand-like tactility. ZONZ™ comfort fit technology uses selected yarns and varying stitch designs to tailor overall glove fit to enhance hand movement for higher dexterity and reduced fatigue. Its silicone-free design is paint- and finish-process-friendly and dirt-masking palm allows longer wear times.
Certifications: ANSI Cut Level A2. ANSI Abrasion Level 4. CE marked. EN388:2003 - 4342. EN 388:2016 - 4X42B. EN 1149. REACH Compliant.
Packaging: Packaged 12 pairs in a bag; 12 bags in a carton.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty