Would you like to log in again?
Nurses Bandage Scissors, OR Grade, Sklar
Supplier: Sklar
11-1496 11-1495
89025-476EA 103.75 USD InStock
89025-476 89025-478
Nurses Bandage Scissors, OR Grade, Sklar
Scissors
Nurses Bandage Scissors are non-ratcheted, finger ring scissors used for removing bandages.
They feature a plastic safety guard attached to the bottom portion of the lower blade, that have a flattened blunt nodule at the tip intended to slide between bandages and skin without harming the skin. The shanks are straight but after the screw lock the blades angle slightly.
The finger rings and safety guard are made of blue latex-free plastic.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty