Would you like to log in again?
Datalogging Heat Stress Monitor, Sper Scientific
Supplier: Sper Scientific
800035
10025-394EA 234.13 USD InStock
10025-394
Datalogging Heat Stress Monitor, Sper Scientific
Data Loggers
Heat stress monitor features a large display that continually monitors all heat stress indicators.
  • Large display that continually monitors temperature, relative humidity, and all heat stress indicators
  • Audible temperature alarm, and records up to 32000 data points that can be downloaded using the USB cable
These include Air Temperature, Relative Humidity, Heat Index, Dew Point, and Wet Bulb. The unit recalls min./max. temperature and provides visual alarms for user-selectable hi/lo temperature. This unit also features an audible temperature alarm, and records up to 32000 data points that can be downloaded using the USB cable and software that is supplied with the unit.
Certifications: CE, RoHS certified.
Ordering information: Includes four AAA batteries, stand for a desk-top or wall mount, USB cable, and software. Unit also includes a five-year manufacturer's warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty