Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TWSG1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-11606R-CY3
10265-644EA 811.1 USD InStock
10265-644
Anti-TWSG1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Twisted Gastrulation Protein 1(TWSG1) is a secreted BMP binding protein structurally related to the BMP antagonists Chordin and Noggin. TSG can inhibit BMP activity by binding directly to BMP proteins thereby preventing BMPs from binding to their receptors. It can act either as a BMP4 agonist or antagonist (depending on the specific biochemical environment) by binding to the BMP4/Chordin complex.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TWSG1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty