Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RBM15 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-12402R-A647
10281-396EA 811.1 USD InStock
10281-396
Anti-RBM15 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
RBM15 is a 977 amino acid protein that localizes to the nucleus and contains one SPOC domain and three RRM domains. Expressed as multiple alternatively spliced isoforms, RBM15 interacts with Epstein-Barr (EBV) viral proteins and is thought to be involved in the regulation of Hox genes, possibly via interactions with RNA and spliceosome components. RBM15 is subject to post-translational phosphorylation, probably by ATM or ATR. Chromosomal aberrations involving the RBM15 gene, which localizes to human chromosome 1, may be associated with the development of acute megakaryoblastic leukemia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RBM15
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty