Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ADAR Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-2168R-CY3
10325-160EA 811.1 USD InStock
10325-160
Anti-ADAR Rabbit Polyclonal Antibody (Cy3®)
Antibodies
ADAR1 converts adenosine to inosine in dsRNA, which destabilizes the dsRNA helix. This activity is important for various functions like site-specific RNA editing of transcripts of the glutamate receptors and modifying viral RNA genomes (which may be responsible for hypermutation of certain negative-stranded viruses, e.g., measles virus). ADAR1 also binds to short interfering RNAs (siRNA) without editing them and suppresses siRNA-mediated RNA interference. This protein is ubiquitously expressed, with the highest levels being found in brain and lung.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADAR
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty