Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NS5A Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-4856R-A647
10399-134EA 811.1 USD InStock
10399-134
Anti-NS5A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
HCV is classified into the genus Hepacivirus of the family Flaviviridae. Like all the members of the family, HCV is an enveloped, single-stranded, positive-sense RNA virus. Its genome (about 9600 nt) is flanked at both termini by conserved, highly structured non-translated regions (NTRs) and encodes a polyprotein precursor (about 3000 aa), which is proteolytically processed by host and viral proteases to produce the structural (core, E1, E2 and p7) and non-structural (NS2, NS3, NS4A, NS4B, NS5A and NS5B) proteins of the virus. Recently, an additional protein has been identified, whose function remains unknown. NS5A is a ~56 kDa pleiotropic protein with key roles in both viral RNA replication and modulation of the physiology of the host cell. It's exact role is not currently known (2008).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NS5A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty