Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DDX58 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-5300R-A350
10412-810EA 811.12 USD InStock
10412-810
Anti-DDX58 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
The innate immune system detects viral infection by recognizing various viral components and triggers antiviral responses. Like the toll-like receptor 3 (TLR3), the cytoplasmic helicase retinoic acid inducible gene protein 1 (RIG1/DDX58) recognizes double-stranded (ds) RNA, a molecular pattern associated with viral infection. Unlike TLR3 however, RIG1/DDX58 activates the kinases TBK1 and IKKe through the adaptor protein IPS1. These kinases then phosphorylate the transcription factors IRF3 and IRF7 which are essential for the expression of type-I interferons. RIG1/DDX58 is required for the production of interferons in response to RNA viruses including paramyxoviruses, influenza virus, and Japanese encephalitis virus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DDX58
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty