Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LCMV Protein Z Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-6946R-CY5.5
10458-416EA 792.04 USD InStock
10458-416
Anti-LCMV Protein Z Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Plays a crucial role in virion assembly and budding. Expressed late in the virus life cycle, it acts as an inhibitor of viral transcription and RNA synthesis by interacting with the viral polymerase L (By similarity). Presumably recruits the NP encapsidated genome to cellular membranes at budding sites via direct interaction with NP. Plays critical roles in the final steps of viral release by interacting with host TSG101, a member of the vacuolar protein-sorting pathway and using other cellular host proteins involved in vesicle formation pathway. The budding of the virus progeny occurs after association of protein Z with the viral glycoprotein complex SSP-GP1-GP2 at the cell periphery, step that requires myristoylation of protein Z. Also selectively represses protein production by associating with host eIF4E

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LCMV Protein Z
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty