Would you like to log in again?
Human Recombinant IP-10 (from E. coli)
Supplier: PeproTech
300-12-25UG 300-12-250UG 300-12-500UG 300-12-100UG 300-12-1MG 300-12-5UG
10773-560EA 1511.41 USD InStock
10773-560 10773-558 10773-556 10773-564 10773-554 10773-562
Human Recombinant IP-10 (from E. coli)
Proteins and Peptides
IP-10 is a CXC chemokine that signals through the CXCR3 receptor. IP-10 selectively chemoattracts Th1 lymphocytes and monocytes, and inhibits cytokine-stimulated hematopoietic progenitor cell proliferation. Additionally, it is angiostatic and mitogenic for vascular smooth muscle cells. Recombinant Human IP-10 (CXCL10) is an 8.6 kDa protein consisting of 77 amino acids, including the four conserved cysteine residues present in CXC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty